Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

2-Nitrobenzaldehyde ≥98%

Glass bottle for solids, 500 g
Catalog # A11501.36
Irritant
2-Nitrobenzaldehyde ≥98%
Catalog # A11501.36
CAS Number:  552-89-6
2-Nitrobenzaldehyde ≥98%
2-Nitrobenzaldehyde ≥98%

Glass bottle for solids, 500 g

CAS Number:  552-89-6
Price
498,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 552-89-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Flash Point: 110°(230°F) Melting Point: 43-46° Boiling Point: 153°/23mm Beilstein Reference: 742624 Merck Reference: 14,6587 MDL No.: MFCD00007132

  • CAS Number:552-89-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007132
  • Molecular Weight:151.12 g/mol
  • Merck Index:14,6587
  • Boiling Pt:153 °C (23 mmHg)
  • Melting Pt:43…46 °C
  • Density:1,35 g/cm³ (25 °C)
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Kamertemperatuur
  • EINECS:209-025-3
Specifications

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    500 g

Specifications