2-Nitrobenzaldehyde ≥98%
Glass bottle for solids, 25 g
Purity: 98+% CAS No.: 552-89-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Flash Point: 110°(230°F) Melting Point: 43-46° Boiling Point: 153°/23mm Beilstein Reference: 742624 Merck Reference: 14,6587 MDL No.: MFCD00007132
- :552-89-6
- :C₇H₅NO₃
- :MFCD00007132
- :151.12 g/mol
- :14,6587
- :153 °C (23 mmHg)
- :43…46 °C
- :1,35 g/cm³ (25 °C)
- :>110 °C (230 °F)
- :Kamertemperatuur
- :209-025-3
Documents
- Verpakking:Glass bottle for solids
- VPE:25 g