Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
2-Nitrobenzaldehyde ≥98%
2-Nitrobenzaldehyde ≥98%
Glass bottle for solids, 25 g
  A11501.14
2-Nitrobenzaldehyde ≥98%
  A11501.14
 :  552-89-6
2-Nitrobenzaldehyde ≥98%
2-Nitrobenzaldehyde ≥98%
Glass bottle for solids, 25 g
 :  552-89-6
54,00 €
1 * 25 g
Purity: 98+% CAS No.: 552-89-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Flash Point: 110°(230°F) Melting Point: 43-46° Boiling Point: 153°/23mm Beilstein Reference: 742624 Merck Reference: 14,6587 MDL No.: MFCD00007132
  •  :552-89-6
  •  :C₇H₅NO₃
  •  :MFCD00007132
  •  :151.12 g/mol
  •  :14,6587
  •  :153 °C (23 mmHg)
  •  :43…46 °C
  •  :1,35 g/cm³ (25 °C)
  •  :>110 °C (230 °F)
  •  :Kamertemperatuur
  •  :209-025-3
  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.