Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Xyleen ≥97%

Glass bottle, 2.500 mL
Catalogus # L13317.0F
Ontvlambaar
Irriterend
Leverancier:   Thermo Fisher Scientific
Xyleen ≥97%
Catalogus # L13317.0F
CAS-nummer:  1330-20-7
Xyleen ≥97%
Xyleen ≥97%

Glass bottle, 2.500 mL

CAS-nummer:  1330-20-7
Price
84,80 €
Log in voor beschikbaarheid
1 * 2.500 mL
Product Details & Documents

Purity: 97+% CAS No.: 1330-20-7 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 29°(84°F) Refractive Index: 1.4970 Odour: Aromatic Melting Point: -25° Boiling Point: 137-144° Beilstein Reference: 1901563 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00077264

Synoniemen:  Dimethylbenzene
  • CAS-nummer:1330-20-7
  • Formule:C₆H₄(CH₃)₂
  • MDL-nummer:MFCD00077264
  • Molecular Weight:106.17 g/mol
  • Merck index:14,10081
  • Kokend Pt:137…144 °C
  • Smelten Pt:< –25 °C
  • Dichtheid:0,860
  • Flits Pt:29 °C (84 °F)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-535-7
  • UN:1307
  • ADR:3,II
Specificaties

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    2.500 mL

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.