Shaking incubators, multitron standard
Product Details & Documents
For applications in microbiology and cell culture, bacteria, yeasts and fungi that require vigorous agitation for optimal results. Also ideal for cell culture where slow and gentle agitation is required or molecular biology for the preparation of Mini and Maxi- preps or for the expression of recombinant proteins. Thanks to the front opening door which supports the tray, the Multitron Standard is simple to use. It provides easy and secure access to all the flasks, regardless of their positions on the tray. The extracted tray is about 1,30 meters above the ground. The lower unit will not need you to dive inside the chamber to manipulate your samples; Simply pull the tray on the door.
- Three basic configurations where different options of refrigeration and orbit (25 and 50 mm) are available
- Accurate temperature control and two large LEDs for a visual inspection of the device status
- Control all units in a stacked group from any control panel
- Standard Multitron is available with refrigeration, a cooling loop is installed in each unit in the ventilation circuit. The main advantage of this compared to the Peltier technology systems is better cooling performance and lower energy consumption
- The front door opens downwards and can be used as a mounting for the shaker table
Bestelinformatie: Supplied without tray. Universal tray adhesive or completed by clamps, pre- fitted trays pliers, special trays for microplates, stainless steel or plastic clamps, tube racks or microplates are available, please contact us.
Specificaties
- Gewicht:135 kg
- Omschrijving:Multitron Standard with cooling (amplitude 25 mm), single chamber with stand
- b×d×h:1070×860×845 mm
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Single unit, refrigerated, with base
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Triple stacked unit, non-refrigerated, with base
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Triple stacked unit, refrigerated, with base
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Double stacked unit, refrigerated, with base
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 100 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 1000 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 2000 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 25 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 250 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Double stacked unit, non-refrigerated, with base
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 50 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 500 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Single unit, non-refrigerated, with base
Productinformatie
- Catalogus #BeschikbaarheidPrice
- ATW universal platform, with Sticky Stuff® adhesive mat, 870×330 mmATW universal platform, with Sticky Stuff® adhesive mat, 870×330 mmINFO69455Log in voor beschikbaarheid1 * 1 ST
Productinformatie
- Catalogus #BeschikbaarheidPrice
- ATM universal platform, with Sticky Stuff® adhesive mat, large, 850×470 mmATM universal platform, with Sticky Stuff® adhesive mat, large, 850×470 mm444-0396Log in voor beschikbaarheid1 * 1 ST
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 5000 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 4000 ml
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Steel flask clamp, 3000 ml
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



