Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Shaking incubators, multitron standard

Shaking incubators, multitron standard

Multitron Standard (amplitude 50 mm), single chamber with stand, 118 kg, 1070×860×845 mm
Catalogus # 444-4229
Leverancier:   INFORS
Shaking incubators, multitron standard
Catalogus # 444-4229
Leverancier:  INFORS
Shaking incubators, multitron standard
Shaking incubators, multitron standard

Multitron Standard (amplitude 50 mm), single chamber with stand, 118 kg, 1070×860×845 mm

Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

For applications in microbiology and cell culture, bacteria, yeasts and fungi that require vigorous agitation for optimal results. Also ideal for cell culture where slow and gentle agitation is required or molecular biology for the preparation of Mini and Maxi- preps or for the expression of recombinant proteins. Thanks to the front opening door which supports the tray, the Multitron Standard is simple to use. It provides easy and secure access to all the flasks, regardless of their positions on the tray. The extracted tray is about 1,30 meters above the ground. The lower unit will not need you to dive inside the chamber to manipulate your samples; Simply pull the tray on the door.

  • Three basic configurations where different options of refrigeration and orbit (25 and 50 mm) are available
  • Accurate temperature control and two large LEDs for a visual inspection of the device status
  • Control all units in a stacked group from any control panel
  • Standard Multitron is available with refrigeration, a cooling loop is installed in each unit in the ventilation circuit. The main advantage of this compared to the Peltier technology systems is better cooling performance and lower energy consumption
  • The front door opens downwards and can be used as a mounting for the shaker table

Ordering information: Supplied without tray. Universal tray adhesive or completed by clamps, pre- fitted trays pliers, special trays for microplates, stainless steel or plastic clamps, tube racks or microplates are available, please contact us.

Specificaties
  • Gewicht:
    118 kg
  • Omschrijving:
    Multitron Standard (amplitude 50 mm), single chamber with stand
  • b×d×h:
    1070×860×845 mm
Accessories

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Single unit, refrigerated, with base
    Single unit, refrigerated, with base
    INFO29791
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Triple stacked unit, non-refrigerated, with base
    Triple stacked unit, non-refrigerated, with base
    INFO28573
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Triple stacked unit, refrigerated, with base
    Triple stacked unit, refrigerated, with base
    INFO28570
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Double stacked unit, refrigerated, with base
    Double stacked unit, refrigerated, with base
    INFO28621
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 100 ml
    Steel flask clamp, 100 ml
    444-0380
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 1000 ml
    Steel flask clamp, 1000 ml
    444-0381
    Log in voor beschikbaarheid
    1 * 1 ST - 55,00 €

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 2000 ml
    Steel flask clamp, 2000 ml
    444-0382
    Log in voor beschikbaarheid
    1 * 1 ST - 145,00 €

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 25 ml
    Steel flask clamp, 25 ml
    444-0383
    Log in voor beschikbaarheid
    1 * 1 ST - 28,00 €

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 250 ml
    Steel flask clamp, 250 ml
    444-0384
    Log in voor beschikbaarheid
    1 * 1 ST - 57,33 €

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Double stacked unit, non-refrigerated, with base
    Double stacked unit, non-refrigerated, with base
    444-0690
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 50 ml
    Steel flask clamp, 50 ml
    444-0387
    Log in voor beschikbaarheid
    1 * 1 ST - 29,00 €

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 500 ml
    Steel flask clamp, 500 ml
    444-0388
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Single unit, non-refrigerated, with base
    Single unit, non-refrigerated, with base
    INFO62020
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • ATW universal platform, with Sticky Stuff® adhesive mat, 870×330 mm
    ATW universal platform, with Sticky Stuff® adhesive mat, 870×330 mm
    INFO69455
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • ATM universal platform, with Sticky Stuff® adhesive mat, large, 850×470 mm
    ATM universal platform, with Sticky Stuff® adhesive mat, large, 850×470 mm
    444-0396
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 5000 ml
    Steel flask clamp, 5000 ml
    444-0389
    Log in voor beschikbaarheid
    1 * 1 ST

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 4000 ml
    Steel flask clamp, 4000 ml
    444-0386
    Log in voor beschikbaarheid
    1 * 1 ST - 110,00 €

Productinformatie

  • Catalogus #
    Beschikbaarheid
    Price
  • Steel flask clamp, 3000 ml
    Steel flask clamp, 3000 ml
    444-0385
    Log in voor beschikbaarheid
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.