Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Shaking incubators, multitron standard

Shaking incubators, multitron standard

Multitron Standard with cooling (amplitude 50 mm), single chamber with stand, 135 kg, 1070×860×845 mm
Catalog # 444-4226
Supplier:   INFORS
Shaking incubators, multitron standard
Catalog # 444-4226
Supplier:  INFORS
Shaking incubators, multitron standard
Shaking incubators, multitron standard

Multitron Standard with cooling (amplitude 50 mm), single chamber with stand, 135 kg, 1070×860×845 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

For applications in microbiology and cell culture, bacteria, yeasts and fungi that require vigorous agitation for optimal results. Also ideal for cell culture where slow and gentle agitation is required or molecular biology for the preparation of Mini and Maxi- preps or for the expression of recombinant proteins. Thanks to the front opening door which supports the tray, the Multitron Standard is simple to use. It provides easy and secure access to all the flasks, regardless of their positions on the tray. The extracted tray is about 1,30 meters above the ground. The lower unit will not need you to dive inside the chamber to manipulate your samples; Simply pull the tray on the door.

  • Three basic configurations where different options of refrigeration and orbit (25 and 50 mm) are available
  • Accurate temperature control and two large LEDs for a visual inspection of the device status
  • Control all units in a stacked group from any control panel
  • Standard Multitron is available with refrigeration, a cooling loop is installed in each unit in the ventilation circuit. The main advantage of this compared to the Peltier technology systems is better cooling performance and lower energy consumption
  • The front door opens downwards and can be used as a mounting for the shaker table

Ordering information: Supplied without tray. Universal tray adhesive or completed by clamps, pre- fitted trays pliers, special trays for microplates, stainless steel or plastic clamps, tube racks or microplates are available, please contact us.

Specifications
  • Gewicht:
    135 kg
  • Omschrijving:
    Multitron Standard with cooling (amplitude 50 mm), single chamber with stand
  • b×d×h:
    1070×860×845 mm
Accessories

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • INFO29791
    Single unit, refrigerated, with base
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • INFO28573
    Triple stacked unit, non-refrigerated, with base
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • INFO28570
    Triple stacked unit, refrigerated, with base
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • INFO28621
    Double stacked unit, refrigerated, with base
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0380
    Steel flask clamp, 100 ml
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0381
    Steel flask clamp, 1000 ml
    Log in for availability
    1 * 1 ST - 55,00 €

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0382
    Steel flask clamp, 2000 ml
    Log in for availability
    1 * 1 ST - 145,00 €

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0383
    Steel flask clamp, 25 ml
    Log in for availability
    1 * 1 ST - 28,00 €

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0384
    Steel flask clamp, 250 ml
    Log in for availability
    1 * 1 ST - 57,33 €

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0690
    Double stacked unit, non-refrigerated, with base
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0387
    Steel flask clamp, 50 ml
    Log in for availability
    1 * 1 ST - 29,00 €

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0388
    Steel flask clamp, 500 ml
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • INFO62020
    Single unit, non-refrigerated, with base
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • INFO69455
    ATW universal platform, with Sticky Stuff® adhesive mat, 870×330 mm
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0396
    ATM universal platform, with Sticky Stuff® adhesive mat, large, 850×470 mm
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0389
    Steel flask clamp, 5000 ml
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0386
    Steel flask clamp, 4000 ml
    Log in for availability
    1 * 1 ST - 110,00 €

Product Information

  • Catalog #
    Omschrijving
    Availability
    Price
  • 444-0385
    Steel flask clamp, 3000 ml
    Log in for availability
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.