Microtubes, with Thread
Micro tubes with neutral screw cap enclosed, skirted base, 0,5 ml, 5000/case
Catalogus # SARS72.730
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Specificaties
- Verpakt:5000/case
- Capaciteit:0,5 ml
- Omschrijving:Micro tubes with neutral screw cap enclosed, skirted base
- Cat. No.:SARS72.730