Orders LinkContactUsLinkComponent
Microtubes
Microtubes
Tubes with screw cap assembled, sterile, with writing space and graduation, 2,0 ml, 5000/case, 2,0 ml
Catalog # SARS72.694.107
Marketsource
Supplier:   SARSTEDT
Microtubes
Catalog # SARS72.694.107
Supplier:  SARSTEDT
Marketsource
Microtubes
Tubes with screw cap assembled, sterile, with writing space and graduation, 2,0 ml, 5000/case, 2,0 ml
Price
461,01 €
Log in for availability
1 * 5.000 ST
Product Details & Documents
PP, translucent
  • Excellent chemical resistance
  • Excellent temperature resistance
Without or with skirted base.
Documents
Specifications
  • Verpakt:
    5000/case
  • Aanbevolen werkvolume:
    2,0 ml
  • Capaciteit:
    2,0 ml
  • Omschrijving:
    Tubes with screw cap assembled, sterile, with writing space and graduation
Accessories
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.006
    Colour coded inserts, blue
    Log in for availability
    1 * 1.000 ST - 17,11 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.712
    Screw cap with seperate retaining loop, neutral
    Log in for availability
    1 * 5.000 ST - 231,32 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713
    Colour coded inserts, neutral
    Log in for availability
    1 * 5.000 ST - 85,56 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.002
    Colour coded inserts, red
    Log in for availability
    1 * 5.000 ST - 85,56 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.007
    Colour coded inserts, violet
    Log in for availability
    1 * 1.000 ST - 17,11 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.009
    Colour coded inserts, white
    Log in for availability
    1 * 1.000 ST - 17,11 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.010
    Colour coded inserts, black
    Log in for availability
    1 * 5.000 ST - 83,64 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.004
    Colour coded inserts, yellow
    Log in for availability
    1 * 5.000 ST - 85,56 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • SARS65.713.005
    Colour coded inserts, green
    Log in for availability
    1 * 5.000 ST - 85,56 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.