Pipette tips
Type A, 200 µl, +, geel, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, 96/box, 480/inner case, 1920/case
Catalogus # SARS70.760.202
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
- Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
- Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Specificaties
- Verpakt:96/box, 480/inner case, 1920/case
- Steriel:+
- Volume:200 µl
- Seq:02
- Tip stijl:Type A
- Tip kleur:geel
- Voor gebruik met:Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical
Accessories
Productinformatie
- Catalogus #KleurBeschikbaarheidPrice
- Tip system box for pipette tip trays, with hinged lid, emptyTip system box for pipette tip trays, with hinged lid, emptySARS95.1298.001transparantLog in voor beschikbaarheid1 * 1 ST