Orders LinkContactUsLinkComponent
Microtubes, Prepared
Microtubes, Prepared
Microtubes, prepared, coagulation citrate, with attached push cap closure, 0,5 ml, 1000/case
Catalogus # SARS41.1506.002
Marketsource
Leverancier:   SARSTEDT
Microtubes, Prepared
Catalogus # SARS41.1506.002
Leverancier:  SARSTEDT
Marketsource
Microtubes, Prepared
Microtubes, prepared, coagulation citrate, with attached push cap closure, 0,5 ml, 1000/case
Price
137,84 €
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Specificaties
  • Verpakt:
    1000/case
  • Capaciteit:
    0,5 ml
  • Omschrijving:
    Microtubes, prepared, coagulation citrate, with attached push cap closure
  • Seq:
    14