Mailing boxes
Mailing boxes with integral restriants for 1 - 4 containers, 198×107×50 mm
Catalogus # SARS95.901
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
- With printed label
- Lightweight
- Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
Specificaties
- Omschrijving:Mailing boxes with integral restriants for 1 - 4 containers
- Seq:02
- b×d×h:198×107×50 mm