Cell scrapers
Celschraper voor gebruik in grote weefselcultuurkolven, 390 mm
SARS83.1831
Marketsource
: SARSTEDT
Cell scraper with soft, flexible blade for gentle removal of monolayer cultures, minimising cell damage.
- Easy and complete separation of monolayer due to extra thin and flexible blade
- When downward pressure is applied to the handle the blade can remove all cells in its path in one pass
- Sterilised by gamma irradiation, individually wrapped
- Omschrijving:Celschraper voor gebruik in grote weefselcultuurkolven
- Seq:03
- Totale lengte:390 mm