Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Xyleen, extra pure
25 L
  KRAF07048.8500
Xyleen, extra pure
  KRAF07048.8500
 :  1330-20-7
Xyleen, extra pure
Xyleen, extra pure
25 L
 :  1330-20-7
357,00 €
1 * 25 L
This product has been blocked by your organization. Please contact your purchasing department for more information
 :  Dimethylbenzene
  •  :1330-20-7
  •  :C₆H₄(CH₃)₂
  •  :MFCD00077264
  •  :106.17 g/mol
  •  :13,10136
  •  :140 °C (1013 hPa)
  •  :–34 °C
  •  :0,86…0.88 g/cm³ (20 °C)
  •  :25 °C
  •  :Kamertemperatuur
  •  :215-535-7
  •  :1307
  •  :3,III
  • VPE:
    25 L