Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 2% in aqueous solution, extra pure

Natriumhydroxide 2% in aqueous solution, extra pure

Plastic bottle, 20 L
Catalogus # KRAF05085.7020
Corrosief
Leverancier:   Bernd Kraft
Natriumhydroxide 2% in aqueous solution, extra pure
Catalogus # KRAF05085.7020
Leverancier:  Bernd Kraft
CAS-nummer:  1310-73-2
Natriumhydroxide 2% in aqueous solution, extra pure

Plastic bottle, 20 L

CAS-nummer:  1310-73-2
Price
204,00 €
Log in voor beschikbaarheid
1 * 20 L
Product Details & Documents
Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    20 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.