Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 20% in aqueous solution, extra pure

Natriumhydroxide 20% in aqueous solution, extra pure

Plastic bottle, 5 L
Catalogus # KRAF05063.5020
Corrosief
Leverancier:   Bernd Kraft
Natriumhydroxide 20% in aqueous solution, extra pure
Catalogus # KRAF05063.5020
Leverancier:  Bernd Kraft
CAS-nummer:  1310-73-2
Natriumhydroxide 20% in aqueous solution, extra pure

Plastic bottle, 5 L

CAS-nummer:  1310-73-2
Price
134,00 €
Log in voor beschikbaarheid
1 * 5 L
Product Details & Documents
Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kokend Pt:119…135 °C (1013 hPa)
  • Smelten Pt:9…10 °C
  • Dichtheid:1,53 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    5 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.