Labels, hazard warning
Hazard labels: non-ion radiation, PIC 311-TRI 050-B7541, Polyester, zwart op geel, 50×43 mm, 7 labels/card
Catalogus # 116-0183
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: non-ion radiation
- Afmetingen:50×43 mm
- Materiaal:Polyester
- Seq:15
- Model:PIC 311-TRI 050-B7541
- Verpakt:7 labels/card