Orders LinkContactUsLinkComponent
Microtubes, Prepared
Microtubes, Prepared
Microtubes, prepared, serum-gel clotting activator, with attached push cap closure, 1,1 ml, 1000/case
Catalog # SARS41.1500.005
Marketsource
Supplier:   SARSTEDT
Microtubes, Prepared
Catalog # SARS41.1500.005
Supplier:  SARSTEDT
Marketsource
Microtubes, Prepared
Microtubes, prepared, serum-gel clotting activator, with attached push cap closure, 1,1 ml, 1000/case
Price
306,63 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Specifications
  • Verpakt:
    1000/case
  • Capaciteit:
    1,1 ml
  • Omschrijving:
    Microtubes, prepared, serum-gel clotting activator, with attached push cap closure
  • Seq:
    11