Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide 8% in aqueous solution, extra pure

Natriumhydroxide 8% in aqueous solution, extra pure

Plastic bottle, 2,5 L
Catalog # KRAF05197.4000
Corrosive
Supplier:   Bernd Kraft
Natriumhydroxide 8% in aqueous solution, extra pure
Catalog # KRAF05197.4000
Supplier:  Bernd Kraft
CAS Number:  1310-73-2
Natriumhydroxide 8% in aqueous solution, extra pure

Plastic bottle, 2,5 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Verpakking:
    Plastic bottle
  • VPE:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.