Orders LinkContactUsLinkComponent
Rods, AR-Glas®
8±0,20 mm, 1500 mm
Catalogus # SCOR1028131
Leverancier:   Schott AG
Rods, AR-Glas®
Catalogus # SCOR1028131
Leverancier:  Schott AG
Rods, AR-Glas®
8±0,20 mm, 1500 mm
Price
192,00 €
Log in voor beschikbaarheid
1 * 15 kg
Product Details & Documents
Made of AR®-Glas.
  • Soft glass is acid-resistant
  • Has high thermal expansion for pressure melting and for fusing with platinum or dumet and other alloys
Specificaties
  • Ø uitw.:
    8±0,20 mm
  • Lengte:
    1500 mm
  • Seq:
    06