Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Methyleenchloride, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised

Glass bottle with septum cap, 250 mL
  83682.230
 
Methyleenchloride, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
  83682.230
 :  75-09-2
Methyleenchloride, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Methyleenchloride, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised

Glass bottle with septum cap, 250 mL

 :  75-09-2
 
84,40 €
   
1 * 250 mL
 

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

 :  Methylene chloride, DCM
  •  :75-09-2
  •  :CH₂Cl₂
  •  :MFCD00000881
  •  :84.93 g/mol
  •  :12,06140
  •  :39,8 °C (1013 hPa)
  •  :–95 °C
  •  :1,322 g/cm³ (20 °C)
  •  :200-838-9
  •  :1593
  •  :6.1,III
  •  :Filtered through a 0.2 µm filter, packaged under nitrogen.
  •  :Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
  •  :Kamertemperatuur
  •  :01-2119480404-41
 

 

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 10 ppm

 

  • Verpakking:
    Glass bottle with septum cap
  • VPE:
    250 mL