Orders LinkContactUsLinkComponent
Pincetten
Pincetten
Super fine point, straight, 115 mm
Catalogus # 241-0332
Leverancier:   EXCELTA
Pincetten
Catalogus # 241-0332
Leverancier:  EXCELTA
Pincetten
Super fine point, straight, 115 mm
Price
79,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Stainless steel, antimagnetic.
  • Economical special forceps for precision applications
  • Corrosion-resistant
Ideal when working with microelectronic assemblies and in the electronics industry.
Specificaties
  • Omschrijving:
    Super fine point, straight
  • Lengte:
    115 mm
  • Seq:
    03