Orders LinkContactUsLinkComponent
Solid phase microextraction fibers
Kit 1: polyacrylate, PDMS, PDMS, for volatiles and semivolatiles, for autosampler use
Catalogus # VARI391896308
Leverancier:   VARIAN
Solid phase microextraction fibers
Catalogus # VARI391896308
Leverancier:  VARIAN
Solid phase microextraction fibers
Kit 1: polyacrylate, PDMS, PDMS, for volatiles and semivolatiles, for autosampler use
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 3 ST
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specificaties
  • Omschrijving:
    Kit 1: polyacrylate, PDMS, PDMS, for volatiles and semivolatiles, for autosampler use
  • Seq:
    23
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Merlin microseal, SPME replacement seal, 23 gauge
    Merlin microseal, SPME replacement seal, 23 gauge
    VARI392609902
    Log in voor beschikbaarheid
    1 * 1 ST - 416,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • SPME fiber holder for CTC autosampler
    SPME fiber holder for CTC autosampler
    VARISU57347U
    Log in voor beschikbaarheid
    1 * 1 ST
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • SPME stand, 15 ml
    SPME stand, 15 ml
    VARISU57357U
    Log in voor beschikbaarheid
    1 * 1 ST
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • SPME inlet guide for manual injection
    SPME inlet guide for manual injection
    VARISU57356U