Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Analytical HPLC columns, Microsorb™ C18

Analytical HPLC columns, Microsorb™ C18

Microsorb™, Analytisch, C18, 100 Å, 5 µm, 4,60 mm, 150 mm
Catalogus # VARIR0089200D5
Leverancier:   VARIAN
Analytical HPLC columns, Microsorb™ C18
Catalogus # VARIR0089200D5
Leverancier:  VARIAN
Analytical HPLC columns, Microsorb™ C18

Microsorb™, Analytisch, C18, 100 Å, 5 µm, 4,60 mm, 150 mm

Price
958,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Microsorb™ C18 is an octadecylsilane-covered silica that has been bonded using monofunctional bonding agents under optimised conditions. This material is then endcapped with trimethylsilyl groups to minimise nonspecific binding, maximise mass recoveries and improve peak shape.

C18 columns are typically used with water/methanol or water/acetonitrile solvent combinations and are ideal for the separation of small molecules, proteins, or peptides with differing hydrophobicities.

Specificaties
  • Kolomtype:
    Analytisch
  • Ø inw.:
    4,60 mm
  • Partikelgrootte:
    5 µm
  • Poriëngrootte:
    100 Å
  • Omschrijving:
    Microsorb™
  • Lengte:
    150 mm
  • Seq:
    01
  • Fase:
    C18
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.