Orders LinkContactUsLinkComponent
Cellulose, Reference Standards
Cellulose, Reference Standards
Microcrystalline Cellulose, USP reference standard, Vial, 1 g
Catalogus # 1098388.
Leverancier:   USP
Cellulose, Reference Standards
Catalogus # 1098388.
Leverancier:  USP
CAS-nummer:  
Cellulose, Reference Standards
Microcrystalline Cellulose, USP reference standard, Vial, 1 g
CAS-nummer:  
Price
434,00 €
Log in voor beschikbaarheid
1 * 1 g
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certificeringen: US Pharmacopea Reference Standard
Documents
Specificaties
Specificaties
  • Verpakking:
    Vial
  • VPE:
    1 g
  • Standaard:
    USP reference standard
  • Omschrijving:
    Microcrystalline Cellulose
  • Cat. No.:
    1098388.
  • Omschrijving:
    Microcrystalline Cellulose
  • CAS No.:
    9004-34-6
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.