Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mitoxantrone Related Compounds, Reference Standard

Mitoxantrone Related Compounds, Reference Standard

Mitoxantrone System Suitability Mixture, USP reference standard, Vial, 0,3 mg
Catalogus # 1445211.
Leverancier:   USP
Mitoxantrone Related Compounds, Reference Standard
Catalogus # 1445211.
Leverancier:  USP
CAS-nummer:  
Mitoxantrone Related Compounds, Reference Standard

Mitoxantrone System Suitability Mixture, USP reference standard, Vial, 0,3 mg

CAS-nummer:  
Price
1 400,00 €
Log in voor beschikbaarheid
1 * 0,3 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Mitoxantrone System Suitability Mixture: Mixture of 0.2 mg 1-Amino-5,8-dihydroxy-4-({2-[(2-hydroxyethyl)amino]ethyl}amino)anthracene-9,10-dione hydrochloride and 0.1 mg Mitoxantrone Hydrochloride

Certificeringen: US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Vial
  • VPE:
    0,3 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Mitoxantrone System Suitability Mixture
  • UN No.:
    2811
  • Cat. No.:
    1445211.
  • Omschrijving:
    Mitoxantrone

Specificaties

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.