Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fexofenadine, Reference Standards

Fexofenadine, Reference Standards

Fexofenadine Hydrochloride, USP reference standard, Vial, 200 mg
Catalogus # 1270377.
Leverancier:   USP
Fexofenadine, Reference Standards
Catalogus # 1270377.
Leverancier:  USP
CAS-nummer:  
Fexofenadine, Reference Standards

Fexofenadine Hydrochloride, USP reference standard, Vial, 200 mg

CAS-nummer:  
Price
405,00 €
Log in voor beschikbaarheid
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Vial
  • VPE:
    200 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Fexofenadine Hydrochloride
  • Cat. No.:
    1270377.
  • Omschrijving:
    Fexofenadine Hydrochloride
  • Formule:
    C₃₂H₄₀ClNO₄
  • CAS No.:
    153439-40-8

Specificaties

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.