VWR® UltraPure, Buffer, PBS solution (phosphate buffered saline)
Phosphate buffered saline (PBS), 1X solution, Dulbecco's formulation, Plastic bottle, 500 mL
Leverancier: VWR Chemicals
VWR® UltraPure, Buffer, PBS solution (phosphate buffered saline)
Catalogus # K812-500ML
Leverancier: VWR Chemicals
CAS-nummer:
Ga naar:
Product Details & Documents
Dulbecco's formulering zonder calcium en magnesium.
- Vrij van DNase, RNase of protease
137,7 mM sodium chloride, 2,7 mM potassium chloride, 9,55 mM phosphate buffer
pH @ 25 °C 7,1 to 7,7
Specificaties
Specificaties
- Verpakking:Plastic bottle
- VPE:500 mL
- Omschrijving:Phosphate buffered saline (PBS), 1X solution, Dulbecco's formulation