DETERGENT SPECIAAL TICKOPUR R 32 1 * 1 L
Catalogus # BAND828
Leverancier: BANDELIN-ELECTRONIC
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Natriumchloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
VWR®, Laboratoriumflessen, smalle hals, met schroefdop<br />met schroefdop
Leverancier: VWR Collection
Ethanol 70% ≥70% (v/v), TechniSolv®
Leverancier: VWR Chemicals