Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Genistein

Genistein

Glass bottle for solids, 250 mg
Catalogus # CAYM10005167-250
Leverancier:   Cayman Chemical
Genistein
Catalogus # CAYM10005167-250
Leverancier:  Cayman Chemical
CAS-nummer:  446-72-0
Genistein

Glass bottle for solids, 250 mg

CAS-nummer:  446-72-0
Price
145,00 €
Log in voor beschikbaarheid
1 * 250 mg
Product Details & Documents
Synoniemen:  5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
  • CAS-nummer:446-72-0
  • Formule:C₁₅H₁₀O₅
  • MDL-nummer:MFCD00016952
  • Molecular Weight:270.24 g/mol
  • Smelten Pt:297…298 °C
  • EINECS:207-174-9

Documents

Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    250 mg

Specificaties