Genistein
Glass bottle for solids, 100 mg
: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
- :446-72-0
- :C₁₅H₁₀O₅
- :MFCD00016952
- :270.24 g/mol
- :297…298 °C
- :207-174-9
- Verpakking:Glass bottle for solids
- VPE:100 mg