3,4',5-Trimethoxybenzophenone
Glass bottle for solids, 10 mg
Catalogus # CAYM10004185-10
Leverancier: Cayman Chemical
3,4',5-Trimethoxybenzophenone
Catalogus # CAYM10004185-10
Leverancier: Cayman Chemical
CAS-nummer: 94709-12-3
Ga naar:
Product Details & Documents
- CAS-nummer:94709-12-3
- Formule:C₁₆H₁₆O₄
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:10 mg



