Pincetten
Straight, short fine tips, 110 mm
Catalogus # 241-0331
Leverancier: EXCELTA
Ga naar:
Product Details & Documents
Stainless steel, antimagnetic.
- Economical special forceps for precision applications
- Corrosion-resistant
Ideal when working with microelectronic assemblies and in the electronics industry.
Specificaties
- Omschrijving:Straight, short fine tips
- Lengte:110 mm
- Seq:01