Objectives
Plan achromatic objective, Ph 100x/1,25, 0,15 mm
Catalogus # 630-0938
Leverancier: OLYMPUS OPTICAL
Ga naar:
Product Details & Documents
Specificaties
- Vergroting:Ph 100x/1,25
- Omschrijving:Plan achromatic objective
- Seq:13
- Werkafstand:0,15 mm