pH/°C meter, handheld, 205
Product Details & Documents
This one-hand, robust food penetration pH/°C measuring instrument comes with automatic temperature compensation. The robust penetration measuring tip is interchangeable and not affected by dirt and dust thanks to the hole diaphragm. The instrument is especially well suited to pH measurements in semi-solid media such as meat.
- pH tip embedded in unbreakable plastic; tip can be exchanged by user
- Combined penetration tip with temperature probe
- Maintenance-free gel electrolyte
- Backlit, 2-line display
- Audible key feedback
- Automatic full-scale value recognition
- 1, 2 or 3 point calibration possible
We can monitor both pH value and temperature simultaneously on the illuminated display. The automatic final value recognition (auto-hold) records the reading as soon as this is stable. An advisory tone informs when the reading is stable. The storage cap, which is filled with gel electrolyte and attached to the pH probe, is used to store the probe between measurements. The electrolyte gel promotes the durability of the pH probe.
The belt/wall bracket supplied enables the measuring instrument to be stored safely on the wall or on a belt.
: DIN EN 12830.
: Testo 205 is supplied with penetration probe, storage cap, belt/wall holder. The starter kit includes the instrument with penetration probe, storage cap, gel and cal. bottles 250 ml pH 4+7, belt/wall holder and aluminium case.
CE Handheld pH Meter with Temperature Penetration Probe 205 CE pH Temperature Meter Handheld 205 testo-CE-Konformitaet_testo205ph1_0213-2017_D_GB testo-CE-Konformitaet_testo205ph1_0214-2017_D_GB
Specificaties
- Omschrijving:Draagbare pH/°C-meter, met temperatuur/insteeksonde, 205
- Seq:01
- pH - kalibratie:1-, 2- of 3-puntskalibratie mogelijk
- pH - nauwkeurigheid:±0,02
- Bedrijfstemperatuurbereik:0...+50 °C
- Temperatuurbereik:0...+60 °C (Short-term to +80 °C max. 5 min)
- b×d×h:145×38×167 mm
- Temperatuurnauwkeurigheid:±0,4 °C
- Voeding:4x knoopcel LR44
- pH - bereik:0,00...+14,00
- Gewicht:0,135 kg
- Automatische bufferherkenning:Automatische waardeherkenning, volledige schaal
- Cat. No.:TEST0563.2051
- Opslagtemperatuur:−20...+70 °C
- Gebruiksduur accu's:80 h (auto off 10 min)
- Model:205
- pH - resolutie:0,01
- IP-klasse:IP 65
- Aflezing:Met achtergrondverlichting, 2-regelig LCD-scherm
- Temperatuurresolutie:0,1 °C
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Spare pH probe with gel storage cap
Productinformatie
- Catalogus #BeschikbaarheidPrice
- pH-bufferoplossing 7,00 in doseerfles (250 ml), met DAkkS-kalibratiecertificaatpH-bufferoplossing 7,00 in doseerfles (250 ml), met DAkkS-kalibratiecertificaatTEST0554.2063Log in voor beschikbaarheid1 * 250 mL - 21,00 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Opslagdop voor testo 205, met KCI-gelvullingOpslagdop voor testo 205, met KCI-gelvullingTEST0554.2051Log in voor beschikbaarheid1 * 1 ST - 18,00 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- pH-bufferoplossing 4,01 in doseerfles (250 ml), met DAkkS-kalibratiecertificaatpH-bufferoplossing 4,01 in doseerfles (250 ml), met DAkkS-kalibratiecertificaatTEST0554.2061Log in voor beschikbaarheid1 * 250 mL - 21,00 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Button cell batteries, Type LR 44, 1,5 V



