Solid phase extraction cartridges, Resprep®, C18 / C8
Resprep® SPE cartridge, high load, endcapped, Cartridge, C8, 3 ml, 200 mg
Catalogus # REST26037
Leverancier: Restek
Ga naar:
Product Details & Documents
These cartridges are made of glass fiber disks embedded with C18 or C8 bonded silica. These hydrophobic (nonpolar) silica-based adsorbents are ideal to extract hydrophobic analytes from polar matrices, such as water (e.g., pesticides from water).
- All cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted
- Extract semivolatile organic compounds
- Deep-pore design reduces clogging and allows faster flow rates
- Lower cost than PTFE disks
: Meet requirements for EPA Methods 525.1, 506, 550.1, and 549.1.
Specificaties
- Kolomtype:Cartridge
- Capaciteit:3 ml
- Omschrijving:Resprep® SPE cartridge, high load, endcapped
- Seq:02
- Bedgewicht:200 mg
- Fase:C8