Orders LinkContactUsLinkComponent
Flow splitters, post-column, QuickSplit
Flow splitters, post-column, QuickSplit
Binary fixed, post column, 10:1 split ratio
Catalogus # REST25329
Leverancier:   Restek
Flow splitters, post-column, QuickSplit
Catalogus # REST25329
Leverancier:  Restek
Flow splitters, post-column, QuickSplit
Flow splitters, post-column, QuickSplit
Binary fixed, post column, 10:1 split ratio
Price
2 610,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Flow splitters with fluid resistor technology eliminate the adjustments to the capillary tubing for optimising split ratio with wide range of interchangeable resistors.
  • Split ratio not affected by changes in viscosity or pressure
  • For adjustable flow splitter, an adjustable metering valve gives convenient control of split ratio
  • High operating pressure limit: fixed flow splitters (10000 psi) and adjustable flow splitter (5000 psi)
  • Low dead volume - negligible effect on analyte bandwidth
  • Total flow: 0,1 to 5,0 ml/min
Specificaties
  • Omschrijving:
    Binary fixed, post column, 10:1 split ratio
  • Seq:
    02