Precision sampling valves for vials, Mininert®
Replacement septa
Catalogus # REST24906
Leverancier: Restek
Ga naar:
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
- Available for screw cap and crimp top vials
- Threaded flange used to secure tight fit
Specificaties
- Omschrijving:Replacement septa
- Seq:07