Orders LinkContactUsLinkComponent
Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 24 mm/400 thread
Catalogus # REST24904
Leverancier:   Restek
Precision sampling valves for vials, Mininert®
Catalogus # REST24904
Leverancier:  Restek
Precision sampling valves for vials, Mininert®
Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 24 mm/400 thread
Price
278,00 €
Log in voor beschikbaarheid
1 * 12 ST
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
  • Available for screw cap and crimp top vials
  • Threaded flange used to secure tight fit
Specificaties
  • Omschrijving:
    Mininert® precision sampling valves, for screw-thread vials, 24 mm/400 thread
  • Seq:
    05
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Replacement septa
    Replacement septa
    REST24906
    Log in voor beschikbaarheid
    1 * 50 ST - 71,30 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Septum insertion tool
    Septum insertion tool
    REST24907
    Log in voor beschikbaarheid
    1 * 1 ST - 64,70 €