Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Inserts for screw thread step vials

Inserts for screw thread step vials

0,50 ml, Flat bottom, Glas, Neen
Catalog # REST21788
Supplier:   Restek
Inserts for screw thread step vials
Catalog # REST21788
Supplier:  Restek
Inserts for screw thread step vials
Inserts for screw thread step vials

0,50 ml, Flat bottom, Glas, Neen

Price
231,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These inserts can be used with 4 ml WISP 48 screw thread step vials.

Specifications
  • Inclusief dop:
    Neen
  • Capaciteit:
    0,50 ml
  • Materiaal:
    Glas
  • Seq:
    02
  • Bodem:
    Flat bottom