Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Packed column FID replacement jets
Packed column FID replacement jets, with standard, 0,46 mm ID tip
  REST21694
 :   Restek
Packed column FID replacement jets
  REST21694
 :  Restek
Packed column FID replacement jets
Packed column FID replacement jets
Packed column FID replacement jets, with standard, 0,46 mm ID tip
95,40 €
1 * 1 ST
These capillary dedicated FID replacement jet for Agilent 5890/6850/6890 GCs are available in standard, high-temperature or Siltek treated high-performance versions.
  • 0,46 mm ID jets are used for most general-purpose packed column applications
  • 0,76 mm ID jets are used for packings that exhibit high bleed and that frequently clog the tip of smaller 0,46 mm jets
The standard version threads are specially coated for easy installation and removal. Also special processing ensures the highest degree of cleanliness. High-performance version - Siltek treated are extremely inert for use with active compounds.
  • Omschrijving:
    Packed column FID replacement jets, with standard, 0,46 mm ID tip
  • Seq:
    01
Accessories
  •  
    Omschrijving
  • REST22328
    FID jet removal kit
    1 * 1 ST - 77,90 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.