Packed GC columns, 2abc refinery column set
Analytical controls configuration, 2,00 mm, 0,05 m
Catalogus # REST88004-875
Leverancier: Restek
Ga naar:
Product Details & Documents
Special columns for unsaturated light hydrocarbons.
- Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.
Specificaties
- Ø inw.:2,00 mm
- Omschrijving:Analytical controls configuration
- Lengte:0,05 m
- Seq:03