Genistein
Glass bottle for solids, 100 mg
Catalogus # ROTH0716.3
Marketsource
Leverancier: Roth Carl
Ga naar:
Product Details & Documents
Synoniemen: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
- CAS-nummer:446-72-0
- Formule:C₁₅H₁₀O₅
- MDL-nummer:MFCD00016952
- Molecular Weight:270.24 g/mol
- Smelten Pt:297…298 °C
- EINECS:207-174-9
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:100 mg



