Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Eosin B

Glass bottle for solids, 25 g
Catalogus # ROTH0306.2
Marketsource
Leverancier:   Roth Carl
Eosin B
Catalogus # ROTH0306.2
Leverancier:  Roth Carl
Marketsource
CAS-nummer:  548-24-3
Eosin B

Glass bottle for solids, 25 g

CAS-nummer:  548-24-3
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents
Synoniemen:  Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
  • CAS-nummer:548-24-3
  • Formule:C₂₀H₆Br₂N₂Na₂O₉
  • MDL-nummer:MFCD00005041
  • Molecular Weight:624.06 g/mol
  • Merck index:12,03638
  • Smelten Pt:> 310 °C
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:208-943-1
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    25 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.