Xyleen
Glass bottle, 1 L
Catalogus # ROTH9713.1
Marketsource
Leverancier: Roth Carl
Ga naar:
Product Details & Documents
Synoniemen: Dimethylbenzene
- CAS-nummer:1330-20-7
- Formule:C₆H₄(CH₃)₂
- MDL-nummer:MFCD00077264
- Molecular Weight:106.17 g/mol
- Merck index:13,10136
- Kokend Pt:140 °C (1013 hPa)
- Smelten Pt:–34 °C
- Dichtheid:0,86…0.88 g/cm³ (20 °C)
- Flits Pt:25 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:215-535-7
- UN:1307
- ADR:3,III
Specificaties
Specificaties
- Verpakking:Glass bottle
- VPE:1 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



