Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Glass bottle for solids, 250 g
Catalog # ROTH9127.1
Marketsource
Supplier:   Roth Carl
Tween® 20 (Polysorbate)
Catalog # ROTH9127.1
Supplier:  Roth Carl
Marketsource
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)
Glass bottle for solids, 250 g
CAS Number:  9005-64-5
Price
27,90 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1,1 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:500-018-3
Specifications
Specifications
  • Verpakking:
    Glass bottle for solids
  • VPE:
    250 g
Specifications