Orders LinkContactUsLinkComponent
D-(+)-Glucose
Glass bottle for solids, 2,5 kg
  ROTHX997.3
Marketsource
 :   Roth Carl
D-(+)-Glucose
  ROTHX997.3
 :  Roth Carl
Marketsource
 :  50-99-7
D-(+)-Glucose
D-(+)-Glucose
Glass bottle for solids, 2,5 kg
 :  50-99-7
112,50 €
1 * 2,5 kg
 :  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  •  :50-99-7
  •  :C₆H₁₂O₆
  •  :MFCD00148912
  •  :180.16 g/mol
  •  :14,04459
  •  :146 °C
  •  :1,54 g/cm³ (20 °C)
  •  :Kamertemperatuur
  •  :200-075-1
  • Verpakking:
    Glass bottle for solids
  • VPE:
    2,5 kg