Natriumhydroxide 50% in aqueous solution
Glass bottle, 1 L
Catalog # ROTH8655.1
Marketsource
Supplier: Roth Carl
Natriumhydroxide 50% in aqueous solution
Catalog # ROTH8655.1
Supplier: Roth Carl
Marketsource
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:119…135 °C (1013 hPa)
- Melting Pt:9…10 °C
- Density:1,53 g/cm³ (20 °C)
- Storage Temperature:Kamertemperatuur
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Specifications
Specifications
- Verpakking:Glass bottle
- VPE:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



