D-(+)-Malic acid
Glass bottle for solids, 5 g
Catalogus # ROTH8633.2
Marketsource
Leverancier: Roth Carl
Ga naar:
Product Details & Documents
Synoniemen: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
- CAS-nummer:636-61-3
- Formule:C₄H₆O₅
- MDL-nummer:MFCD00004245
- Molecular Weight:134.09 g/mol
- Merck index:13,05307
- Kokend Pt:322 °C (1013 hPa)
- Smelten Pt:101…104 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:211-262-2
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:5 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



