D-(+)-Malic acid
Glass bottle for solids, 1 g
Catalogus # ROTH8633.1
Marketsource
Leverancier: Roth Carl
Ga naar:
Product Details & Documents
Synoniemen: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
- CAS-nummer:636-61-3
- Formule:C₄H₆O₅
- MDL-nummer:MFCD00004245
- Molecular Weight:134.09 g/mol
- Merck index:13,05307
- Kokend Pt:322 °C (1013 hPa)
- Smelten Pt:101…104 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:211-262-2
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:1 g



