Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Koper(II)sulfaat

Koper(II)sulfaat

Glass bottle for solids, 1 kg
Catalog # ROTHP023.2
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Koper(II)sulfaat
Catalog # ROTHP023.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7758-98-7
Koper(II)sulfaat

Glass bottle for solids, 1 kg

CAS Number:  7758-98-7
Price
132,50 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Cupric sulphate
  • CAS Number:7758-98-7
  • Formula:CuSO₄
  • MDL Number:MFCD00010981
  • Molecular Weight:159.61 g/mol
  • Merck Index:12,02722
  • Melting Pt:200 °C
  • Density:3,603 g/cm³ (25 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:231-847-6
  • UN:3288
  • ADR:6.1,III
Specifications

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.